Learn More
Zwiebel™ Class M1 Box Weights with Certificate
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.
Brand:  Zwiebel™ PBI23121
 View more versions of this product
Product Code. 11780763
  • 235.00 DKK / Each
Only null left
Estimated Shipment: 29-07-2025
Description
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
±3 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class M1 Box Weights with Certificate > Mass: 50g