Learn More
Zwiebel™ Class F1 Weight Box with Certificate
Brand:  Zwiebel™ PBI56812
 View more versions of this product
Product Code. 11810892
  • 1295.00 DKK / Each
Only null left
Estimated Shipment: 18-09-2025
Specifications
500 g
±2.50 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class F1 Weight Box with Certificate > Mass: 500g