Learn More
VITLAB™ Class B SAN Tall Form Measuring Cylinders
Brand:  VITLAB™ 65091
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10004510
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance
View alternative products »
Description
Description
  • Crystal clear graduated cylinder
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
Specifications
±2mL
SAN (Styrene-acrylonitrile)
B
250mL
±2mL
315mm
Class B tolerances according to DIN 12681/ISO 6706
47mm
Raised Scale and Ring Marks at the Primary Scale Points
±2mL
Alternative Products