Få mere at vide
VITLAB™ Klasse B polypropylen graduerede cylindre med hævet blå skala
Gradueringer:
0.5 mL
1 mL
5 mL
10 mL
20 mL
Pakningsstørrelse:
Hver
Pakke med 12
Pakke med 6
Beskrivelse
- Highly transparent polypropylene graduated cylinder
- Hexagonal base with bottom studs provides high stability
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
- To preserve markings, do not clean at temperatures exceeding 60°C
Tekniske data
Tekniske data
| Klasse | B |
| Gradueringer | 20 mL |
| Grænse for fejl | ±20mL |
| Form | Round |
| Skala farve | Blue |
| Base Type | Hexagonal |
| Højde (metrisk) | 482 mm |
| Type | Graduated Measuring Cylinder |