Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper
Borosilicate volumetric flask with glass stopper and ISO batch certificate.
Brand:  BRAND™ DAKKS/37266
1427.81 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 17917210
  • 1667.55 DKK / Each
In Stock
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Borosilicate 3.3
  • DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • All BLAUBRAND™ volumetric flasks are delivered with a batch number and batch certificate included in the original packaging. They are also available with individual certificates or DAkkS calibration certificate upon request.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 L
Autoclavable
Yes
Non-sterile
Borosilicate Glass (3.3)
Reusable
Narrow
Flat
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper > Volumetric Flask, 5 L