Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper
Borosilicate volumetric flask with glass stopper and ISO batch certificate.
Brand: BRAND™ DAKKS/37266
1427.81 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate 3.3
- DIN EN ISO 1042
- Calibrated to contain (TC, In)
- All BLAUBRAND™ volumetric flasks are delivered with a batch number and batch certificate included in the original packaging. They are also available with individual certificates or DAkkS calibration certificate upon request.
Specifications
5 L | |
Autoclavable | |
Yes | |
Non-sterile |
Borosilicate Glass (3.3) | |
Reusable | |
Narrow | |
Flat |
Product Content Correction
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper > Volumetric Flask, 5 L
Spot an opportunity for improvement?Share a Content Correction