missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Class 3.1 Independent Temperature Safety Device
For Use With: BINDER units ED 56, ED 115, ED 260, ED 720
Brand: BINDER™ 8012-0985
This item is not returnable.
View return policy
Description
- Class 3.1
- With visual alarm
- Safety devices also available in Class 2 and 3.3
Specifications
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) | |
Plastic | |
Temperature Regulation |
BINDER Chambers ED 56, ED 115, ED 260, ED 720 | |
Temperature Safety Device |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Class 3.1 Independent Temperature Safety Device > For Use With: ED 260, ED 115, ED 720, FD 56, ED 56, FED 115, FD 260, FED 56 models
Spot an opportunity for improvement?Share a Content Correction