Få mere at vide
Clarithromycin-N-methyl-d3, TRC
Artikelnummer. 30357633
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30357633 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30357633 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5597525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clarithromycin-N-methyl-d3
CAS 959119-22-3
Renhedsgrad noter HPLC
Molekylær formel C38 D3 H66 N O13
Mængde 5 mg
Synonym 6-O-Methylerythromycin-d3, 6-O-Methylerythromycin A-d3, A 56268-d3, Abbott 56268-d3, Antibiotic A 56268-d3, Antibiotic TE 31-d3, BIAXIN XL-d3, Biaxin-d3, CRIXAN OD-d3, Clamicin-d3, Claric-d3, Claris-d3, Claritek-d3, Clarith-d3, Clarithro-d3, Clathromycin-d3, Crixan-d3, Fascar-d3, Fromilid-d3, Kelamycin-d3, Klacid-d3, Klaribac-d3, Klaricid-d3, Klarithran MR-d3, Macladin-d3, Naxy-d3, TE 031-d3, Veclam-d3, Zeclar-d3
InChI formel InChI=1S/C38H69NO13/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,47-14)33(52-35-29(41)25(39(11)12)16-20(3)48-35)22(5)30(23(6)34(44)50-26)51-27-18-36(8,46-13)32(43)24(7)49-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1/i11D3
SMIL [2H]C([2H])([2H])N(C)[C@H]1C[C@@H](C)O[C@@H](O[C@@H]2[C@@H](C)[C@H](O[C@H]3C[C@@](C)(OC)[C@@H](O)[C@H](C)O3)[C@@H](C)C(=O)O[C@H](CC)[C@@](C)(O)[C@H](O)[C@@H](C)C(=O)[C@H](C)C[C@@]2(C)OC)[C@@H]1O
IUPAC navn (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-6-[(2S,3R,4S,6R)-3-hydroxy-6-methyl-4-[methyl(trideuteriomethyl)amino]oxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
Molekylvægt (g/mol) 750.9718
Formel vægt 750.4957
Procent renhed >95