Få mere at vide
Clarithromycin-N-methyl-13C, d3, TRC
Artikelnummer. 30360405
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30360405 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360405 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5597541MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Clarithromycin-N-methyl-13C, D3
CAS 2699608-30-3
Renhedsgrad noter HPLC
Molekylær formel 13C C37 H66 D3 N O13
Mængde 1 mg
Synonym 6-O-Methylerythromycin-13C, d3, 6-O-Methylerythromycin A-13C, d3, A 56268-13C, d3, Abbott 56268-13C, d3, Antibiotic A 56268-13C, d3, Antibiotic TE 31-13C, d3, BIAXIN XL-13C, d3, Biaxin-13C, d3, CRIXAN OD-13C, d3, Clamicin-13C, d3, Claric-13C, d3, Claris-d3, Claritek-13C, d3, Clarith-13C, d3, Clarithro-13C, d3, Clathromycin-13C, d3, Crixan-13C, d3, Fascar-13C, d3, Fromilid-13C, d3, Kelamycin-13C, d3, Klacid-13C, d3, Klaribac-13C, d3, Klaricid-13C, d3, Klarithran eclar-13C, d3
InChI formel InChI=1S/C38H69NO13/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,47-14)33(52-35-29(41)25(39(11)12)16-20(3)48-35)22(5)30(23(6)34(44)50-26)51-27-18-36(8,46-13)32(43)24(7)49-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1/i11+1D3
SMIL [2H][13C]([2H])([2H])N(C)[C@H]1C[C@@H](C)O[C@@H](O[C@@H]2[C@@H](C)[C@H](O[C@H]3C[C@@](C)(OC)[C@@H](O)[C@H](C)O3)[C@@H](C)C(=O)O[C@H](CC)[C@@](C)(O)[C@H](O)[C@@H](C)C(=O)[C@H](C)C[C@@]2(C)OC)[C@@H]1O
IUPAC navn (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-6-[(2S,3R,4S,6R)-3-hydroxy-6-methyl-4-[methyl(trideuterio(1^{13}C)methyl)amino]tetrahydropyran-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
Molekylvægt (g/mol) 751.9645
Formel vægt 751.4991
Procent renhed >95