missing translation for 'onlineSavingsMsg'
Få mere at vide

Clarithromycin, 98%, BLD Pharm™

Artikelnummer. 31449398
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
5 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31449398 500 g
31435024 5 g
31442874 100 mg
31448386 100 g
31447152 1 kg
31440868 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31449398 Leverandør BLD Pharm Leverandørnr. BD121855500g
 Begrænset vare
Forventet forsendelse: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Clarithromycin
CAS 81103-11-9
Molekylær formel C38H69NO13
MDL nummer MFCD00865140
Mængde 500 g
SMIL C[C@@](C[C@@H](C)C([C@@H]1C)=O)([C@H](O[C@@]([H])([C@@H]2O)O[C@@H](C[C@@H]2N(C)C)C)[C@@H](C)[C@H](O[C@@]([H])(C[C@]3(OC)C)O[C@H]([C@@H]3O)C)[C@@H](C)C(O[C@@H]([C@@](O)([C@@H]1O)C)CC)=O)OC
Molekylvægt (g/mol) 747.95
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.