Få mere at vide
Cladinose, TRC
Artikelnummer. 30368744
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368744 10 mg
30367168 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368744 Leverandør Toronto Research Chemicals Leverandørnr. TRCC55748010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 470-12-2
Molekylær formel C8H16O4
Molekylvægt (g/mol) 176.21
Synonym 2,6-Dideoxy-3-C-methyl-3-O-methyl-ribo-hexose, Azithromycin Impurity 4, (4R,5S,6S)-4-Methoxy-4,6-dimethyltetrahydro-2H-pyran-2,5-diol
IUPAC navn (3S,4R,5R)-4,5-dihydroxy-3-methoxy-3-methylhexanal
SMIL C[C@H]([C@H]([C@](C)(CC=O)OC)O)O
Tekniske data
Kemisk navn eller materiale Cladinose
Mængde 10 mg
InChI formel InChI=1S/C8H16O4/c1-6(10)7(11)8(2,12-3)4-5-9/h5-7,10-11H,4H2,1-3H3/t6-,7-,8+/m1/s1
Formel vægt 176.1049