missing translation for 'onlineSavingsMsg'
Få mere at vide

Abnova™ CKM Recombinant Protein

Artikelnummer. p-3718133
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16133241

Brand: Abnova™ H00001158P01.10ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Human CKM full-length ORF recombinant protein with GST-tag at N-terminal

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family.

  • Theoretical MW (kDa): 67.65
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MPFGNTHNKFKLNYKPEEEYPDLSKHNNHMAKVLTLELYKKLRDKETPSGFTVDDVIQTGVDNPGHPFIMTVGCVAGDEESYEVFKELFDPIISDCHGGYKPTDKHKTDLNHENLKGGDDLDPNYVLSSRVRTGRSIKGYTLPPHCSRGERRAVEKLSVEALNSLAGEFKGKYYPLKSMTEKEQQQLIDDHFLFDKPVSPLLLASGMARDWPDARGIWHNDNKSLLVWVNEEDHLRVISMEKGGNMKEVFRRFCVGLQKIEEIFKKAGHPFMWNQHLGYVLTCPSNLGTGLRGGVHVKLAHLSKHPKFEEILTRLRLQKRGTGGVDTAAVGSVFDVSNADRLGSSEVEQVQLVVDGVKLMVEMEKKLEKGQSIDDMIPAQK

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Tekniske data

Adgangsnummer AAH07462
Til brug med (applikation) Antibody Production, Protein Array, ELISA, Western Blot
Formulering 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gen-id (Entrez) 1158
Molekylvægt (g/mol) 67.7
Navn CKM (Human) Recombinant Protein (P01)
pH-område 8
Fremstillingsmetode In vitro wheat germ expression system
Oprensningsmetode Glutathione Sepharose 4 Fast Flow
Kvalitetskontrol test 12.5% SDS-PAGE Stained with Coomassie Blue.
Mængde 10 μg
Kilde Wheat Germ (in vitro)
Immunogen MPFGNTHNKFKLNYKPEEEYPDLSKHNNHMAKVLTLELYKKLRDKETPSGFTVDDVIQTGVDNPGHPFIMTVGCVAGDEESYEVFKELFDPIISDCHGGYKPTDKHKTDLNHENLKGGDDLDPNYVLSSRVRTGRSIKGYTLPPHCSRGERRAVEKLSVEALNSLAGEFKGKYYPLKSMTEKEQQQLIDDHFLFDKPVSPLLLASGMARDWPDARGIWHNDNKSLLVWVNEEDHLRVISMEKGGNMKEVFRRFCVGLQKIEEIFKKAGHPFMWNQHLGYVLTCPSNLGTGLRGGVHVKLAHLSKHPKFEEILTRLRLQKRGTGGVDTAAVGSVFDVSNADRLGSSEVEQVQLVVDGVKLMVEMEKKLEKGQSIDDMIPAQK
Opbevaringskrav Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CKMM/M-CK
Fælles navn CKM
Gen symbol CKM
Krydsreaktivitet Human
Arter Wheat Germ (in vitro)
Rekombinant Recombinant
Protein tag GST
Form Solution
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.