missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ CK1 gamma-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586401
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Purity is > 95% by SDS-PAGE.
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
Tekniske data
| CK1 gamma-2 | |
| Polyclonal | |
| Unconjugated | |
| Csnk1g2 | |
| 2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103236, 1455, 65278 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78368, Q62762, Q8BVP5 | |
| Csnk1g2 | |
| Synthetic peptide, corresponding to the N-terminal of Human Casein Kinase Igamma2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion