Learn More
Citric Acid Monohydrate, Supelco™
Product Code. 15893840
Change view
Click to view available options
Quantity:
5 kg
25 kg
Unit Size:
25kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15893840 5 kg 5kg
15856830 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15893840 Supplier Supelco Supplier No. 1002445000
 Restricted Item
Forventet forsendelse: 18-11-2026
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5949-29-1
Specifications
Melting Point 135°C to 152°C
Density 1.54 g/cm3 at 20°C
Quantity 5 kg
Molecular Formula HOC(COOH)(CH2COOH)2 · H2O
MDL Number MFCD00149972
Molecular Weight (g/mol) 210.14
Grade Ungraded
Chemical Name or Material Citric Acid Monohydrate