missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Citric Acid, Monohydrate Granular/ACS), Macron Fine Chemicals™
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Citric Acid, Monohydrate |
| CAS | 5949-29-1 |
| Molekylær formel | C6H10O8 |
| Synonym | citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp |
| InChI nøgle | YASYEJJMZJALEJ-UHFFFAOYSA-N |
| SMIL | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
| IUPAC navn | 2-hydroxypropan-1,2,3-tricarboxylsyre;hydrat |
| Molekylvægt (g/mol) | 210.138 |
| PubChem CID | 22230 |
| ChEBI | CHEBI:31404 |
| Show More |
Safety and Handling
Product Identifier
- Citric Acid Monohydrate
- Advarsel
- Hudætsning/hudirritation Kategori 2
- H319-Forårsager alvorlig øjenirritation.
- P264-Vask grundigt efter brug.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- P305+P351+P338-VED KONTAKT MED ØJNENE: Skyl forsigtigt med vand i flere minutter. Fjern eventuelle kontaktlinser, hvis dette kan gøres let. Fortsæt skylning.
- P337+P313-Ved vedvarende øjenirritation: Søg lægehjælp.
- MIXTURE LIST-Contain: Citric acid, monohydrate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?