Få mere at vide
Citric Acid-2,2,4,4-d4, TRC
Artikelnummer. 30351566
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30351566 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351566 Leverandør Toronto Research Chemicals Leverandørnr. TRCC52100225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Citric Acid-2,2,4,4-d4
CAS 147664-83-3
Renhedsgrad noter HPLC
Molekylær formel C6 2H4 H4 O7
Mængde 25 mg
Synonym 1,2,3-Propane-1,1,3,3-d4-tricarboxylic acid, 2-hydroxy- (9CI), 2-Hydroxy-1,2,3-propane-1,1,3,3-d4-tricarboxylic acid, Citric acid-d4, Citric-2,2,4,4-d4 acid, Citric-2,2,4,4-D4 acid, Citric acid-2,2,4,4-d4
InChI formel InChI=1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1D2,2D2
SMIL [2H]C([2H])(C(=O)O)C(O)(C(=O)O)C([2H])([2H])C(=O)O
IUPAC navn 1,1,3,3-tetradeuterio-2-hydroxypropane-1,2,3-tricarboxylic acid
Molekylvægt (g/mol) 196.15
Formel vægt 196.0521
Procent renhed >95