Få mere at vide
Citrate-dextrose Solution (ACD), TRC
Artikelnummer. 30354416
Skift visning
Mængde:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354416 50 mL
30353927 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354416 Leverandør Toronto Research Chemicals Leverandørnr. TRCC52051550ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 8013-89-6
Molekylær formel C12H20O13
Molekylvægt (g/mol) 372.28
Synonym Acid Citrate-dextrose, Anticoagulant Citrate-dextrose, ACD Solution, Glugicir
IUPAC navn 2-hydroxypropane-1,2,3-tricarboxylic acid;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMIL FC1=C(F)C(N)=C(F)C(N)=C1F
Tekniske data
Kemisk navn eller materiale Citrate-dextrose Solution (ACD)
Mængde 50 mL
InChI formel InChI=1S/C6H8O7.C6H12O6/c7-3(8)1-6(13,5(11)12)2-4(9)10;7-1-3(9)5(11)6(12)4(10)2-8/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1,3-6,8-12H,2H2/t;3-,4+,5+,6+/m.0/s1