Få mere at vide
Citalopram-d6, TRC
Artikelnummer. 30367886
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367886 10 mg
30353926 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367886 Leverandør Toronto Research Chemicals Leverandørnr. TRCC50500210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Citalopram-d6
CAS 1190003-26-9
Renhedsgrad noter HPLC
Molekylær formel C20 2H6 H15 F N2 O
Mængde 10 mg
Synonym Citalopram-D6 Hydrobromide
InChI formel InChI=1S/C20H21FN2O/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20/h4-9,12H,3,10-11,14H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])N(CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3)C([2H])([2H])[2H]
IUPAC navn 1-[3-[bis(trideuteriomethyl)amino]propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile
Molekylvægt (g/mol) 330.43
Formel vægt 330.2015
Procent renhed >95