Få mere at vide
Citalopram-d6 Oxalate, TRC
Artikelnummer. 30358957
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30358957 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358957 Leverandør Toronto Research Chemicals Leverandørnr. TRCC50500310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Citalopram-d6 Oxalate
CAS 1246819-94-2
Molekylær formel C22H17D6FN2O5
Mængde 10 mg
Synonym 1-[3-(Dimethylamino-d6)propyl]-1-(4-fluorophenyl)-1,3-dizohydro-5-isobenzofurancarbonitrile Oxalate Salt
InChI formel InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3)C([2H])([2H])[2H].OC(=O)C(=O)O
IUPAC navn 1-[3-[bis(trideuteriomethyl)amino]propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
Molekylvægt (g/mol) 420.46
Formel vægt 420.1968