Få mere at vide
(±)-Citalopram-d4 HBr (4-fluorophenyl-d4), CDN
Artikelnummer. 30317234
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317234 0.005 g
30319762 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317234 Leverandør CDN Leverandørnr. CDND68220.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Citalopram-d4 hydrobromide
CAS 1219803-58-3
Molekylær formel C20 2H4 H17 F N2 O . Br H
Mængde 0.005 g
InChI formel InChI=1 S/C20H21FN2O.BrH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12 H,3,10-11,14H2,1-2H3;1 H/i5D,6 D,7 D,8 D;
SMIL Br.[2 H]c1c([2 H])c(c([2 H])c([2 H])c1F)C2(CCCN(C)C)OCc3cc(ccc23)C#N
IUPAC navn 1-[3-(dimethylamino)propyl]-1-(2,3,5,6-tetradeuterio-4-fluorophenyl)-3 H-2-benzofuran-5-carbonitrile;hydrobromide
Molekylvægt (g/mol) 409.33
Formel vægt 408.1151 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity