missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CISH 293T Cell Overexpression Lysate (Denatured), Abnova
Beskrivelse
Western Blotting
Tekniske data
Tekniske data
| Cellelinje | 293T |
| Til brug med (applikation) | Western Blot |
| Oversigt | CISH 293T Cell Transient Overexpression Lysate(Denatured) |
| Genadgangsnr. | NM_145071 |
| Molekylvægt (g/mol) | 28.7 |
| pH-område | 6.8 |
| Plasmid | pCMV-CISH full-length |
| Kvalitetskontrol test | Transient overexpression cell Lysate was tested with Anti-CISH antibody (H00001154-B01) by western blotting |
| Mængde | 100 μL |
| Opbevaringsbuffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL CISH 293T Cell Transient Overexpression Lysa
- Advarsel
- Hudsensibilisering Kategori 1
- H317-Kan forårsage allergisk hudreaktion.
- P261-Undgå indånding af pulver/røg/gas/tåge/damp/spray.
- P272-Tilsmudset arbejdstøj bør ikke fjernes fra arbejdspladsen.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?