Få mere at vide
Cisapride-13C,d3, TRC
Artikelnummer. 30350202
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350202 1 mg
30362949 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350202 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4964021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1285970-69-5
Molekylær formel C2213CH26D3ClFN3O4
Molekylvægt (g/mol) 469.96
Synonym rel-4-Amino-5-chloro-N-[1-[(3R,4S)-3-(4-fluorophenoxy)propyl]-3-(methoxy-13C,d3)-4-piperidinyl]-2-methoxybenzamide, (+/-)-Cisapride-13C,d3, Acenalin-13C,d3, Alimix-13C,d3, Cipril-13C,d3, Coordinax-13C,d3, Prepulsid-13C,d3, Propulsin-13C,d3, cis-2-(Methoxy-13C,d3)-4-amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]benzamide
IUPAC navn 4-amino-5-chloro-N-[(3S,4R)-1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl]-2-(trideuterio(113C)methoxy)benzamide
SMIL FC1=CC=C(OCCCN2C[C@H](OC)[C@H](NC(C3=C(O[13C]([2H])([2H])[2H])C=C(N)C(Cl)=C3)=O)CC2)C=C1
Tekniske data
Kemisk navn eller materiale Cisapride-13C,d3
Mængde 1 mg
InChI formel InChI=1S/C23H29ClFN3O4/c1-30-21-13-19(26)18(24)12-17(21)23(29)27-20-8-10-28(14-22(20)31-2)9-3-11-32-16-6-4-15(25)5-7-16/h4-7,12-13,20,22H,3,8-11,14,26H2,1-2H3,(H,27,29)/t20-,22+/m1/s1/i1+1D3