Få mere at vide
cis,trans-Sorbic Acid, TRC
Artikelnummer. 30006637
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006637 25 mg
30008877 50 mg
30009851 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006637 Leverandør Toronto Research Chemicals Leverandørnr. TRCS67689825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5309-57-9
Molekylær formel C6H8O2
Molekylvægt (g/mol) 112.13
Synonym (2Z,4E)-2,4-Hexadienoic acid, 2-cis,4-trans-Hexadienoic acid
IUPAC navn (2Z,4E)-hexa-2,4-dienoic acid
SMIL C\C=C\C=C/C(=O)O
Tekniske data
Kemisk navn eller materiale cis,trans-Sorbic Acid
Mængde 25 mg
InChI formel InChI=1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4-
Formel vægt 112.0524