Få mere at vide
cis-trans-10,11-Epoxy Farnesenic Acid-d3 Methyl Ester (>85%), TRC
Artikelnummer. 30376100
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30376100 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376100 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5894175MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale cis-trans-10,11-Epoxy Farnesenic Acid-d3 Methyl Ester
CAS 951116-90-8
Molekylær formel C16H23D3O3
Mængde 5 mg
Synonym (2Z,6E)-9-(3,3-Dimethyl-2-oxiranyl)-3,7-dimethyl-2,6-nonadienoic Acid Methyl-d3 Ester
InChI formel InChI=1S/C16H26O3/c1-12(9-10-14-16(3,4)19-14)7-6-8-13(2)11-15(17)18-5/h7,11,14H,6,8-10H2,1-5H3/b12-7+,13-11-/i5D3
SMIL [2H]C([2H])([2H])OC(=O)\C=C(\C)/CC\C=C(/C)\CCC1OC1(C)C
Molekylvægt (g/mol) 269.39
Formel vægt 269.207