Få mere at vide
cis-Terbinafine Hydrochloride, TRC
Artikelnummer. 30134989
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134989 100 mg
30136922 25 mg
30005049 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134989 Leverandør Toronto Research Chemicals Leverandørnr. TRCT107490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 176168-78-8
Molekylær formel C21 H25 N . Cl H
Molekylvægt (g/mol) 327.89
Synonym 1-Naphthalenemethanamine, N-[(2Z)-6,6-dimethyl-2-hepten-4-yn-1-yl]-N-methyl-, hydrochloride (1:1), 1-Naphthalenemethanamine, N-(6,6-dimethyl-2-hepten-4-ynyl)-N-methyl-, hydrochloride, (Z)-, 1-Naphthalenemethanamine, N-[(2Z)-6,6-dimethyl-2-hepten-4-ynyl]-N-methyl-, hydrochloride (9CI), (2Z)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. B (EP) as Hydrochloride, cis-Terbinafine Hydrochloride
IUPAC navn (Z)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMIL Cl.CN(C\C=C/C#CC(C)(C)C)Cc1cccc2ccccc12
Tekniske data
Kemisk navn eller materiale cis-Terbinafine Hydrochloride
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C21H25N.ClH/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19;/h5-7,9-14H,16-17H2,1-4H3;1H/b9-5-;
Formel vægt 327.1754
Procent renhed >95%