Få mere at vide
Tocris Bioscience™ cis MZ 1
Negative Control for MZ1
Brand: Tocris Bioscience™ 6155/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Negative Control for MZ 1 (Cat. No. 6154)
cis MZ 1 is a negative Control for MZ1 (Cat.No. 6154). Exhibits no significant VHL binding affinity.Tekniske data
| PROTAC Controls | |
| 1797406-72-4 | |
| 1002.64g/mol | |
| Bromodomain Inhibitors |
| Bromodomain | |
| C49H60ClN9O8S2 | |
| Store at -20°C | |
| 5 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only