Få mere at vide
Tocris Bioscience™ cis MZ 1
GREENER_CHOICE
Negative Control for MZ1
Brand:  Tocris Bioscience™ 6155/5
Encompass
Produktkode 15747236
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Negative Control for MZ 1 (Cat. No. 6154)
cis MZ 1 is a negative Control for MZ1 (Cat.No. 6154). Exhibits no significant VHL binding affinity.
Tekniske data
Tekniske data
PROTAC Controls
1797406-72-4
1002.64g/mol
Bromodomain Inhibitors
Bromodomain
C49H60ClN9O8S2
Store at -20°C
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only