Få mere at vide
cis-Linalool Oxide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | cis-Linalool Oxide |
| CAS | 5989-33-3 |
| Renhedsgrad noter | GC |
| Molekylær formel | C10H18O2 |
| Mængde | 10 mg |
| Synonym | cis-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, Tetrahydro-α,α,5-trimethyl-5-vinyl-furfuryl Alcohol, rel-(2R,5S)-5-Ethenyltetrahydro-α,α,5-trimethyl-2-furanmethanol, (Z)-Furanoid Linalool Oxide, (Z)-Linalool Oxide B, Epoxydihydrolinalool II, Linalool B Oxide, Linalool Oxide, Linalool Oxide 1, Linalool Oxide B, Linalool Oxide I, cis-2-Methyl-2-vinyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, cis-2-Vinyl-2-methyl-5-(1-hydroxy-1-methylethyl)tetrahydrofuran, cis-Furan Linalool Oxide, cis-Furanoid Linalool Oxide, cis-Linalool 3,6-Oxide, cis-Linalool Oxide (furan), cis-Linalool Oxide (furanoid), cis-f-Linalool Oxide, cis-α,α,5-Trimethyl-5-vinyltetrahydrofurfuryl Alcohol |
| InChI formel | InChI=1S/C10H18O2/c1-5-10(4)7-6-8(12-10)9(2,3)11/h5,8,11H,1,6-7H2,2-4H3/t8-,10-/m1/s1 |
| SMIL | C=C[C@]1(O[C@@H](C(C)(O)C)CC1)C |
| IUPAC navn | 2-[(2R,5S)-5-ethenyl-5-methyloxolan-2-yl]propan-2-ol |
| Molekylvægt (g/mol) | 170.25 |
| Vis mere |