Få mere at vide
cis-L-3-Hydroxyproline, TRC
Artikelnummer. 17948004
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17948004 25 mg
17907344 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17948004 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95290025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 567-35-1
Molekylær formel C5H9NO3
Molekylvægt (g/mol) 131.13
Synonym (3R)-3-Hydroxy-L-proline, (2S,3R)-3-Hydroxypyrrolidine-2-carboxylic Acid, (2S,3R)-3-Hydroxypyrrolidine-2-carboxylic Acid, 3-Allohydroxy-L-proline, cis-3-Hydroxy-L-proline, cis-3-Hydroxyproline;
IUPAC navn (2S,3R)-3-hydroxypyrrolidine-2-carboxylic acid
SMIL O[C@@H]1CCN[C@@H]1C(=O)O
Tekniske data
Kemisk navn eller materiale cis-L-3-Hydroxyproline
InChI formel InChI=1S/C5H9NO3/c7-3-1-2-6-4(3)5(8)9/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1
Mængde 25 mg
Formel vægt 131.0582