Få mere at vide
cis-ent-Tadalafil-d3, TRC
Artikelnummer. 30426282
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30426282 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426282 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0045321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale cis-ent-Tadalafil-d3
CAS 1329799-70-3
Molekylær formel C22H16D3N3O4
Mængde 1 mg
Synonym cis-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(methyl-d3)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, rel-(6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-(methyl-d3)pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, (±)-cis-Tadalafil-d3, (±)-cis-Tadalafil-D3 (N-methyl-D3)
InChI formel InChI=1S/C22H19N3O4/c1-24-10-19(26)25-16(22(24)27)9-14-13-4-2-3-5-15(13)23-20(14)21(25)12-6-7-17-18(8-12)29-11-28-17/h2-8,16,21,23H,9-11H2,1H3/t16-,21+/m1/s1/i1D3
SMIL O=C1N(C([2H])([2H])[2H])CC(N2[C@@H](C3=CC=C(OCO4)C4=C3)C5=C(C(C=CC=C6)=C6N5)C[C@@]21[H])=O
IUPAC navn (6S,12aR)-6-(benzo[d][1,3]dioxol-5-yl)-2-(methyl-d3)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
Molekylvægt (g/mol) 392.42
Formel vægt 392.16