Få mere at vide
(+)-cis-Diltiazem-d3 HCl (acetoxy-d3), TRC
Artikelnummer. 30377784
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30377784 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377784 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4606215MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+)-cis-Diltiazem-d3 HCl (acetoxy-d3)
CAS 1217860-13-3
Molekylær formel C22 D3 H23 N2 O4 S . Cl H
Mængde 5 mg
Synonym (2S,3S)-3-(Acetyloxy-d3)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride (1:1), Diltiazem-d3 hydrochloride, (+)-cis-Diltiazem-d3 hydrochloride, Diltiazem-D3 (acetyl-D3) hydrochloride, (+)-cis-Diltiazem-d3 HCl (acetoxy-d3)
InChI formel InChI=1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1/i1D3;
SMIL Cl.[2H]C([2H])([2H])C(=O)O[C@@H]1[C@@H](Sc2ccccc2N(CCN(C)C)C1=O)c3ccc(OC)cc3
IUPAC navn [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] 2,2,2-trideuterioacetate;hydrochloride
Molekylvægt (g/mol) 453.9973
Formel vægt 453.1568