Få mere at vide
cis-Diammine-diiodo Platinum II, TRC
Artikelnummer. 30361400
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361400 500 mg
30354977 250 mg
30360388 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30361400 Leverandør Toronto Research Chemicals Leverandørnr. TRCD416260500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13841-96-8
Molekylær formel H6I2N2Pt
Molekylvægt (g/mol) 482.95
Synonym Diamminediiodo-platinum, Diamminediiodoplatinum
SMIL I[Pt-2]([NH3+])([NH3+])I
Tekniske data
Kemisk navn eller materiale cis-Diammine-diiodo Platinum II