Få mere at vide
cis-5-Tetradecenoic Acid, TRC
Artikelnummer. 17949634
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17949634 10 mg
17948574 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17949634 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29160010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5684-70-8
Molekylær formel C14 H26 O2
Molekylvægt (g/mol) 226.36
Synonym cis-δ5-Tetradecenoic Acid, (Z)-5-Tetradecenoic Acid, C14:1w9, (5Z)-5-Tetradecenoic Acid
IUPAC navn (Z)-tetradec-5-enoic acid
SMIL CCCCCCCC\C=C/CCCC(=O)O
Tekniske data
Kemisk navn eller materiale cis-5-Tetradecenoic Acid
Mængde 10 mg
InChI formel InChI=1S/C14H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h9-10H,2-8,11-13H2,1H3,(H,15,16)/b10-9-
Formel vægt 226.1933