Få mere at vide
cis-5,11-Diethyl-5,6,11,12-tetrahydro-2,8-chrysenediol, TRC
Artikelnummer. 30378550
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378550 50 mg
30372611 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378550 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44507050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 138090-06-9
Molekylær formel C22H24O2
Molekylvægt (g/mol) 320.42
Synonym (5R,11R)-rel-5,11-Diethyl-5,6,11,12-tetrahydro-2,8-chrysenediol, 5,11-cis-Diethyl-5,6,11,12-tetrahydrochrysene-2,8-diol, cis-THC
SMIL CC[C@@H]1Cc2cc(O)ccc2C3=C1c4ccc(O)cc4C[C@H]3CC
Tekniske data
Kemisk navn eller materiale cis-5,11-Diethyl-5,6,11,12-tetrahydro-2,8-chrysenediol
Mængde 50 mg
InChI formel InChI=1S/C22H24O2/c1-3-13-9-15-11-17(23)6-8-20(15)22-14(4-2)10-16-12-18(24)5-7-19(16)21(13)22/h5-8,11-14,23-24H,3-4,9-10H2,1-2H3/t13-,14-/m1/s1
Formel vægt 320.1776