Få mere at vide
cis-4-Decen-1-ol, TRC
Artikelnummer. 30353683
Skift visning
Mængde:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353683 5 g
30358716 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30353683 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2256305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57074-37-0
Molekylær formel C10H20O
Molekylvægt (g/mol) 156.27
Synonym (Z)-4-Decen-1-ol, (4Z)-4-Decen-1-ol, cis-4-Decenol, (Z)-Dec-4-en-1-ol
IUPAC navn (Z)-dec-4-en-1-ol
SMIL CCCCC\C=C/CCCO
Tekniske data
Kemisk navn eller materiale cis-4-Decen-1-ol
Mængde 5 g
InChI formel InChI=1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h6-7,11H,2-5,8-10H2,1H3/b7-6-
Formel vægt 156.1514