Få mere at vide
cis-4-(1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl)phenol, TRC
Artikelnummer. 30422762
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422762 50 mg
30423189 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30422762 Leverandør Toronto Research Chemicals Leverandørnr. TRCT29386550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale cis-4-(1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl)phenol
CAS 14089-22-6
Molekylær formel C23H22O2
Mængde 50 mg
Synonym cis-4-(6-Methoxy-2-phenyl-1,2,3,4-tetrahydro-1-naphthalenyl)phenol, rel-4-[(1R,2S)-1,2,3,4-Tetrahydro-6-methoxy-2-phenyl-1-naphthalenyl]phenol
InChI formel InChI=1S/C23H22O2/c1-25-20-12-14-22-18(15-20)9-13-21(16-5-3-2-4-6-16)23(22)17-7-10-19(24)11-8-17/h2-8,10-12,14-15,21,23-24H,9,13H2,1H3/t21-,23+/m1/s1
SMIL COC1=CC2=C(C=C1)[C@H]([C@H](CC2)C3=CC=CC=C3)C4=CC=C(C=C4)O
IUPAC navn 4-[(1R,2S)-6-methoxy-2-phenyl-1,2,3,4-tetrahydronaphthalen-1-yl]phenol
Molekylvægt (g/mol) 330.42
Formel vægt 330.162