Learn More
cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer
Product Code. 11484754
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484754 25 g 25g
11474754 5 g 5g
11494754 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11484754 Supplier Thermo Scientific Alfa Aesar Supplier No. A13734.14
 Restricted Item
På lager
Add to basket
CAS: 928-94-9 | C6H12O | 100.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 928-94-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063209
InChI Key ZCHHRLHTBGRGOT-PLNGDYQASA-N
Synonym cis-2-hexen-1-ol, 2-hexen-1-ol, z, cis-hex-2-en-1-ol, z-2-hexen-1-ol, z-hex-2-en-1-ol, unii-871tel510e, 2z-hexen-1-ol, 2z-hex-2-en-1-ol, cis-1-hydroxy-2-hexene, cis-2-hexenol
PubChem CID 5324489
IUPAC Name (Z)-hex-2-en-1-ol
SMILES CCC\C=C/CO
Specifications
Density 0.847
Boiling Point 158°C
Flash Point 54°C (129°F)
Refractive Index 1.441
Quantity 25 g
UN Number UN1987
Beilstein 1719708
Formula Weight 100.16
Percent Purity 94%
Assay remainder mainly trans isomer
Chemical Name or Material cis-2-Hexen-1-ol