Få mere at vide
cis-[2-Bromomethyl-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl Benzoate, TRC
Artikelnummer. 17997934
Skift visning
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17997934 1 g
17999664 5 g
17958434 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17997934 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6854051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61397-56-6
Molekylær formel C18 H15 Br Cl2 O4
Molekylvægt (g/mol) 446.12
Synonym (2R,4R)-rel-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol 4-Benzoate, 1,3-Dioxolane-4-methanol, 2-(bromomethyl)-2-(2,4-dichlorophenyl)-, benzoate, (2R,4R)-rel- (9CI), 1,3-Dioxolane-4-methanol, 2-(bromomethyl)-2-(2,4-dichlorophenyl)-, benzoate, cis-, (2R,4R)-rel-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolane-4-methanol benzoate, cis-[2-Bromomethyl-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl benzoate, [(2RS,4RS)-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl Benzoate, [(2RS,4RS)-2-(Bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl Benzoate
IUPAC navn [(2R,4R)-2-(bromomethyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl benzoate
SMIL Clc1ccc(c(Cl)c1)[C@@]2(CBr)OC[C@H](COC(=O)c3ccccc3)O2
Tekniske data
Kemisk navn eller materiale cis-[2-Bromomethyl-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl]methyl Benzoate
Mængde 1 g
InChI formel InChI=1S/C18H15BrCl2O4/c19-11-18(15-7-6-13(20)8-16(15)21)24-10-14(25-18)9-23-17(22)12-4-2-1-3-5-12/h1-8,14H,9-11H2/t14-,18-/m0/s1
Formel vægt 443.9531