Få mere at vide
cis-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, TRC
Artikelnummer. 30003836
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30003836 250 mg -
30009691 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003836 Leverandør Toronto Research Chemicals Leverandørnr. TRCD435930250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61397-58-8
Molekylær formel C14 H14 Cl2 N2 O3
Molekylvægt (g/mol) 329.18
Synonym (2R,4S)-rel-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, cis-, rel-(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol, [(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methanol
IUPAC navn ((2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl)methanol
SMIL OC[C@H]1CO[C@@](Cn2ccnc2)(O1)c3ccc(Cl)cc3Cl
Tekniske data
Kemisk navn eller materiale ((2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1-Himidazol-1-ylmethyl)-1,3-dioxolan-4-yl)methanol
Mængde 250 mg
Renhedsgrad noter HPLC
InChI formel InChI=1S/C14H14Cl2N2O3/c15-10-1-2-12(13(16)5-10)14(8-18-4-3-17-9-18)20-7-11(6-19)21-14/h1-5,9,11,19H,6-8H2/t11-,14-/m0/s1
Formel vægt 328.0381
Procent renhed >95%