Få mere at vide
cis-[2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl p-Tolylsulfonate, TRC
Artikelnummer. 30138724
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138724 1 g
17931935 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138724 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4357181G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 134071-44-6
Molekylær formel C21 H20 Cl2 N2 O5 S
Molekylvægt (g/mol) 483.36
Synonym 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-(4-methylbenzenesulfonate), (2R,4R)-rel-, 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-methylbenzenesulfonate (ester), (2R,4R)-rel- (9CI), 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-methylbenzenesulfonate (ester), cis-, Ketoconazole Imp. E (EP)
IUPAC navn [(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
SMIL Cc1ccc(cc1)S(=O)(=O)OC[C@H]2CO[C@@](Cn3ccnc3)(O2)c4ccc(Cl)cc4Cl
Tekniske data
Kemisk navn eller materiale cis-[2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl p-Tolylsulfonate
Mængde 1 g
InChI formel InChI=1S/C21H20Cl2N2O5S/c1-15-2-5-18(6-3-15)31(26,27)29-12-17-11-28-21(30-17,13-25-9-8-24-14-25)19-7-4-16(22)10-20(19)23/h2-10,14,17H,11-13H2,1H3/t17-,21+/m1/s1
Formel vægt 482.047