Få mere at vide
cis-13-Docosenamide (>85%), TRC
Artikelnummer. 30010907
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010907 5 g
30005128 10 g
30010764 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010907 Leverandør Toronto Research Chemicals Leverandørnr. TRCP5795035G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 112-84-5
Molekylær formel C22 H43 N O
Molekylvægt (g/mol) 337.58
Synonym Plastic additive 21, Erucamide R, (13Z)-13-Docosenamide, (Z)-13-Docosenamide, Erucamide, Adogen 58, Alflow 10, Alflow P 10, Amide E, Ampacet 10090, Armid E, Armoslip E, Armoslip EPX, Armoslip EXP, Atmer 1753, Atmer SA 1753, Atmer SA 1753FD, CESA PPA 0050079, Chemstat HTSA 22, Chemstat HTSA 22-20M, Crodamide E, Crodamide ER, Diamid L 200, E 10, E 10 (amide), Erucic acid amide, Erucic amide, Erucoyl amide, Erucyl amide, Fatty Amide E, Finawax E, Finawax ER, Kemamide E, Kemamide E Ultra, Kemamide E Ultrabead, L 300, L 300 (amide), Loxamid E, Loxiol E SPEZ P, Lutamide ER, MB-S 1519, Neutron S, NewS, PEX-SLT 01, Pinnacle 2530, Proaid AC 18E, RX 4515, SA 1753FD, Struktol TR 131, TEP BP-SL 1, Unislip 1753, WK 1890, cis-13-Docosenamide
IUPAC navn (Z)-docos-13-enamide
SMIL CCCCCCCC\C=C/CCCCCCCCCCCC(=O)N
Tekniske data
Kemisk navn eller materiale Plastic additive 21
Mængde 5 g
InChI formel InChI=1S/C22H43NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H2,23,24)/b10-9-
Formel vægt 337.3345