Få mere at vide
cis-1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6, TRC
Artikelnummer. 30427312
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30427312 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427312 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2942411MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale cis-1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6
CAS 203578-24-9
Molekylær formel C8 D6 H3 N O2
Mængde 1 mg
Synonym cis-1,2,3,6-Tetrahydrophthalimide D6 (3,3,4,5,6,6-D6), 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4','3a,4,7,7a-tetrahydro-4,7-d2-','(3aR,7aS)-rel-, 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4','3a,4,7,7a-tetrahydro-4,7-d2-','cis- (ZCI)
InChI formel InChI=1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)/t5-,6+/i1D,2D,3D2,4D2
SMIL [2H]C1=C([2H])C([2H])([2H])[C@H]2[C@H](C(=O)NC2=O)C1([2H])[2H]
IUPAC navn rac-(3aR,7aS)-4,4,5,6,7,7-hexadeuterio-3a,7a-dihydroisoindole-1,3-dione
Molekylvægt (g/mol) 157.2
Formel vægt 157.101