missing translation for 'onlineSavingsMsg'
Learn More

Ciproxifan Maleate, TRC

Product Code. 30367078
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30367078 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367078 Supplier Toronto Research Chemicals Supplier No. TRCC48400025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Ciproxifan Maleate
CAS 184025-19-2
Molekylær formel C16 H18 N2 O2 . C4 H4 O4
Mængde 25 mg
Synonym Cyclopropyl[4-[3-(1H-imidazol-4-yl)propoxy]phenyl]-Methanone (2Z)-2-Butenedioate, FUB359 Maleate
InChI formel InChI=1S/C16H18N2O2.C4H4O4/c19-16(12-3-4-12)13-5-7-15(8-6-13)20-9-1-2-14-10-17-11-18-14;5-3(6)1-2-4(7)8/h5-8,10-12H,1-4,9H2,(H,17,18);1-2H,(H,5,6)(H,7,8)/b;2-1-
SMIL OC(=O)\C=C/C(=O)O.O=C(C1CC1)c2ccc(OCCCc3c[nH]cn3)cc2
IUPAC navn (Z)-but-2-enedioic acid;cyclopropyl-[4-[3-(1H-imidazol-4-yl)propoxy]phenyl]methanone
Molekylvægt (g/mol) 386.4
Formel vægt 386.1478
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.