missing translation for 'onlineSavingsMsg'
Få mere at vide

Ciprofloxacin monohydrochloride, MedChemExpress

Artikelnummer. 16442149
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16442149 1 g 1g
16400898 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16442149 Leverandør Medchem Express Leverandørnr. HYB0356A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Ciprofloxacin (Bay-09867) monohydrochloride is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin monohydrochloride induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin monohydrochloride has anti-proliferative activity and induces apoptosis. Ciprofloxacin monohydrochloride is a fluoroquinolone antibiotic, exhibiting potent antibacterial activity.

Ciprofloxacin (Bay-09867) monohydrochloride is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin monohydrochloride induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin monohydrochloride has anti-proliferative activity and induces apoptosis. Ciprofloxacin monohydrochloride is a fluoroquinolone antibiotic, exhibiting potent antibacterial activity.

Tekniske data

Kemisk navn eller materiale Ciprofloxacin monohydrochloride
Fysisk form Solid
CAS 93107-08-5
Farve White
Mængde 1 g
Renhedsgrad noter Research
Molekylær formel C17H19ClFN3O3
Til brug med (applikation) COVID-19-immunoregulation
Synonym Bay-09867 monohydrochloride
Opløselighedsinformation H2O : 12.5 mg/mL (33.99 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (13.59 mM; Need ultrasonic)
SMIL [H]Cl.O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CCNCC4)=C3)C1=O)O
Molekylvægt (g/mol) 367.8
Formel vægt 367.8
Procent renhed 98.15%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.