Få mere at vide
Ciprofloxacin-d8 Hydrochloride, TRC
Artikelnummer. 30349771
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30349771 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349771 Leverandør Toronto Research Chemicals Leverandørnr. TRCC48250250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ciprofloxacin D8 hydrochloride
CAS 1216659-54-9
Renhedsgrad noter HPLC
Molekylær formel C17 2H8 H10 F N3 O3 . Cl H
Mængde 50 mg
Synonym 1-Cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid hydrochloride, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl-2,2,3,3,5,5,6,6-d8)-3-quinolinecarboxylic acid hydrochloride (1:1), Ciprofloxacin-d8 hydrochloride, Ciprofloxacin-D8 (piperazine-D8) hydrochloride
InChI formel InChI=1S/C17H18FN3O3.ClH/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;/h7-10,19H,1-6H2,(H,23,24);1H/i3D2,4D2,5D2,6D2;
SMIL Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4)C1([2H])[2H]
IUPAC navn 1-cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molekylvægt (g/mol) 375.85
Formel vægt 375.1601
Procent renhed >95