Få mere at vide
Cipargamin, MedChemExpress
Artikelnummer. 16498667
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
2mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16498667 5 mg 5mg
16440216 2 mg 2mg
16436466 100 mg -
16436068 10 mg -
16422207 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16498667 Leverandør Medchem Express Leverandørnr. HY14430
MedChemExpress Cipargamin (NITD609) is an potent antimalarial compound, with an IC50 of appr 1 nM against P. falciparum.
Cipargamin (NITD609) is an potent antimalarial compound, with an IC50 of appr 1 nM against P. falciparum.
Tekniske data
Kemisk navn eller materiale Cipargamin
Fysisk form Solid
CAS 1193314-23-6
Farve Brown
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C19H14Cl2FN3O
Til brug med (applikation) COVID-19-immunoregulation
Synonym NITD609 KAE609
Opløselighedsinformation DMSO : 50 mg/mL (128.13 mM; Need ultrasonic)
SMIL FC1=C(Cl)C=C(NC2=C3C[C@H](C)N[C@@]2(C4=C(C=CC(Cl)=C4)N5)C5=O)C3=C1
Molekylvægt (g/mol) 390.24
Formel vægt 390.24
Procent renhed 98.3%