Få mere at vide
Cinnarizine EP Impurity E, Mikromol™
Artikelnummer. 30462187
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462187 4 x 25 mg
30049880 25 mg
17992066 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462187 Leverandør Mikromol Leverandørnr. MM1038.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cinnarizine EP Impurity E, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Bis(diphenylmethyl)piperazine
CAS 216581-01-0
Analyt- eller komponentnavne 1,4-Bis(diphenylmethyl)piperazine
Forsendelsestilstand Room Temperature
Molekylær formel C30 H30 N2
Certificeringer/compliance ISO 17025
Synonym 1,4-Bis(diphenylmethyl)piperazine, Cinnarizine Imp. E (EP), Cinnarizine Impurity E
InChI formel InChI=1S/C30H30N2/c1-5-13-25(14-6-1)29(26-15-7-2-8-16-26)31-21-23-32(24-22-31)30(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-20,29-30H,21-24H2
Løsningstype Neat
SMIL C1CN(CCN1C(c2ccccc2)c3ccccc3)C(c4ccccc4)c5ccccc5
IUPAC navn 1,4-dibenzhydrylpiperazine
Molekylvægt (g/mol) 418.57
Mængde 4 x 25 mg
Formel vægt 418.2409 g/mol