missing translation for 'onlineSavingsMsg'
Learn More

Cinnamylideneacetophenone, 98+%, Thermo Scientific Chemicals

Catalog Number 11492068
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
614-57-3
C17H14O
234.298
MFCD00020692
QONKLJMPKWQQFG-HPIZBCMHSA-N
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
1549519
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2
This item is not returnable. View return policy
614-57-3
C17H14O
234.298
MFCD00020692
QONKLJMPKWQQFG-HPIZBCMHSA-N
cinnamylideneacetophenone, cinnamalacetophenone, 5-phenyl-2,4-pentadienophenone, 5-phenylpenta-2,4-dienophenone, 2e,4e-1,5-diphenylpenta-2,4-dien-1-one, ?,e-1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadienophenone, 5-phenyl, 2,4-pentadien-1-one, 1,5-diphenyl-, ?,e, 1,5-diphenyl-2,4-pentadien-1-one, 2,4-pentadien-1-one, 1,5-diphenyl
1549519
(2E,4E)-1,5-diphenylpenta-2,4-dien-1-one
C1=CC=C(C=C1)C=CC=CC(=O)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to 102°C
Quantity 100 g
Beilstein 1870052
Formula Weight 234.3
Percent Purity ≥98%
Chemical Name or Material Cinnamylideneacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'