Få mere at vide
Tocris Bioscience™ Cinacalcet hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand:  Tocris Bioscience™ 6170/10
Encompass
Produktkode 15797236
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.
Tekniske data
Tekniske data
Calcium-sensing Receptor (CaSR) Allosteric Agonist, also CYP2D2 Inhibitor and Ca2+ Channel Blocker
364782-34-3
393.87g/mol
Calcium-Sensing Receptor Agonists
Calcium-Sensing Receptor
C22H22F3N.HCl
Store at -20°C
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Produkttitel
Vælg et problem
For Research Use Only