Learn More
Cinacalcet Hydrochloride, TRC
Product Code. 30137809
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137809 100 mg 100mg
30136238 50 mg 50mg
17911345 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137809 Supplier Toronto Research Chemicals Supplier No. TRCC441800100MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 364782-34-3
Molecular Formula C22 H22 F3 N . Cl H
Molecular Weight (g/mol) 393.87
Synonym 1-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (αR)-, 1-Naphthalenemethanamine, α-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride, (αR)- (9CI), AMG 073 HCl, Cinacalcet hydrochloride, KRN 1493, Mimpara, N-[(1R)-1-(1-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride, Regpara, Sensipar, [(1R)-1-(Naphthalen-1-yl)ethyl]([3-[3-(trifluoromethyl)phenyl]propyl])amine hydrochloride
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride
SMILES Cl.C[C@@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m1./s1
Formula Weight 393.1471
Percent Purity >95%
Chemical Name or Material Cinacalcet Hydrochloride
Grade HPLC