Få mere at vide
Tocris Bioscience™ Cinacalcet hydrochloride
Beskrivelse
Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.
Tekniske data
Tekniske data
| CAS | 364782-34-3 |
| Mængde | 10 mg |
| Mål | Calcium-Sensing Receptor |
| Molekylær formel | C22H22F3N.HCl |
| Formel vægt | 393.87g/mol |
| Produkttype | Inhibitor |
For Research Use Only