Få mere at vide
Tocris Bioscience™ Cinacalcet hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 6170/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.Tekniske data
| Calcium-sensing Receptor (CaSR) Allosteric Agonist, also CYP2D2 Inhibitor and Ca2+ Channel Blocker | |
| 364782-34-3 | |
| 393.87g/mol | |
| Calcium-Sensing Receptor Agonists |
| Calcium-Sensing Receptor | |
| C22H22F3N.HCl | |
| Store at -20°C | |
| 50 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only